trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number A13171.22
also known as A13171-22
Price (USD)/ Each
182.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
182.00
Add to cart
trans-2-Nitrocinnamic acid, 98%, Thermo Scientific Chemicals
Catalog numberA13171.22
Price (USD)/ Each
182.00
-
Add to cart
Chemical Identifiers
CAS1013-96-3
IUPAC Name(2E)-3-(2-nitrophenyl)prop-2-enoic acid
Molecular FormulaC9H7NO4
InChI KeyBBQDLDVSEDAYAA-AATRIKPKSA-N
SMILESOC(=O)C=CC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (HPLC)≥97.5%
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
Melting Point (clear melt)243.0-249.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only