Thermo Scientific Chemicals

2-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A13180.30
250 g, Each
Thermo Scientific Chemicals

2-Aminobenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: A13180.30
250 g, Each
Quantity
Catalog number: A13180.30
also known as A13180-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5344-90-1
IUPAC Name
(2-aminophenyl)methanol
Molecular Formula
C7H9NO
InChI Key
VYFOAVADNIHPTR-UHFFFAOYSA-N
SMILES
NC1=CC=CC=C1CO
Identification (FTIR)
Conforms
Melting Point
81.0-86.0°C
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream to yellow to brown
Assay (GC)
≥97.5%

Description

2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.

Solubility
Very soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text