Thermo Scientific Chemicals

Cyclohexene oxide, 98+%, Thermo Scientific Chemicals

Catalog number: A13185.36
500 g, Each
Thermo Scientific Chemicals

Cyclohexene oxide, 98+%, Thermo Scientific Chemicals

Catalog number: A13185.36
500 g, Each
Quantity
Catalog number: A13185.36
also known as A13185-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
286-20-4
Assay (GC)
≥98.0%
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4505-1.4545 @ 20?C

Description

It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, dyestuffs and in agrochemical industries. It is used in medicine. Cyclohexene oxide (epoxycyclohexane) is an useful monomer in polymerization and coating industry. It is used in the synthesis of alicyclic target materials including pharmaceuticals, perfumery and dyestuffs. It is used as a monomer in photopolymerizations, with carbonmonoxide to yield aromatic polycarbonates which has minimum impurities.

Solubility
Insoluble in water

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text