Thermo Scientific Chemicals

4-Methylcyclohexanone, 98+%, Thermo Scientific Chemicals

Catalog number: A13186.22
100 g, Each
Thermo Scientific Chemicals

4-Methylcyclohexanone, 98+%, Thermo Scientific Chemicals

Catalog number: A13186.22
100 g, Each
Quantity
Catalog number: A13186.22
also known as A13186-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
589-92-4
IUPAC Name
4-methylcyclohexan-1-one
Molecular Formula
C7H12O
InChI Key
VGVHNLRUAMRIEW-UHFFFAOYSA-N
SMILES
CC1CCC(=O)CC1
Refractive Index
1.4440-1.4480 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Form
Liquid
Identification (FTIR)
Conforms

Description

4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylcyclohexanone was used in the synthesis of the pinguisane-type sesquiterpenoid acutifolone A, in racemic form. It is used as intermediates of CCNU and it can be also be used as a solvent.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text