Thermo Scientific Chemicals

BAPTA, 97%, Thermo Scientific Chemicals

Catalog number: A13190.03
1 g, Each
Thermo Scientific Chemicals

BAPTA, 97%, Thermo Scientific Chemicals

Catalog number: A13190.03
1 g, Each
Quantity
Catalog number: A13190.03
also known as A13190-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85233-19-8
IUPAC Name
tetrapotassium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Molecular Formula
C22H20K4N2O10
InChI Key
MAWIDOKQOLSOFT-UHFFFAOYSA-J
SMILES
[K+].[K+].[K+].[K+].[O-]C(=O)CN(CC([O-])=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC([O-])=O)CC([O-])=O
Melting Point
ca 165-175°C dec. (non-U.S. sourced material)
Appearance (Color)
White to pale cream to pale yellow
Comment
Material sourced in the U.S. and in other countries
Form
Powder
Assay (HPLC)
≥96.0%

Description

Acts as a calcium chelatorBAPTA is used as a selective calcium chelating reagent. It is suitable for the spectrophotometric monitoring of calcium levels. It is a lipophilic diesters of the chelating agent used for the inhibition of proteolytic activities of certain metalloproteinases, calpain and transcatheter arterial chemoembolization (TACE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acts as a calcium chelatorBAPTA is used as a selective calcium chelating reagent. It is suitable for the spectrophotometric monitoring of calcium levels. It is a lipophilic diesters of the chelating agent used for the inhibition of proteolytic activities of certain metalloproteinases, calpain and transcatheter arterial chemoembolization (TACE).

Solubility
Soluble in water and sodium bicarbonate.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text