Thermo Scientific Chemicals

Tetramethylethylene, 97%, Thermo Scientific Chemicals

Catalog number: A13201.14
25 g, Each
Thermo Scientific Chemicals

Tetramethylethylene, 97%, Thermo Scientific Chemicals

Catalog number: A13201.14
25 g, Each
Quantity
Catalog number: A13201.14
also known as A13201-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
563-79-1
IUPAC Name
2,3-dimethylbut-2-ene
Molecular Formula
C6H12
InChI Key
WGLLSSPDPJPLOR-UHFFFAOYSA-N
SMILES
CC(C)=C(C)C
Form
Liquid
Appearance (Color)
Clear colorless
Refractive Index
1.4100-1.4150 @ 20?C (UK sourced material only)
Assay (GC)
≥96.0%

Description

Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.

Solubility
Soluble in water (0.071 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text