Propionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propionitrile, act as solvents and are reacted further for various application including, Extraction solvent for fatty acids, oils and unsaturated hydrocarbons, Solvent for spinning and casting, removing agent of colouring matters and aromatic alcohols, Non-aqueous solvent for titrations and for inorganic salts, Recrystallization of steroids, Parent compound for organic synthesis, chemical intermediate in biochemistry, Catalyst and component of transition-metal complex catalysts, Stabilizer for chlorinated solvents, Chemical intermediate and solvent for perfumes and pharmaceuticals. It is used as a dielectric fluid.
Solubility
It decomposes with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Take measures to prevent the build up of electrostatic charge. Incompatible with Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
RUO – Research Use Only
General References:
- R. G. Lerner.; B. P. Dailey. Microwave Spectrum and Structure of Propionitrile. J. Chem. Phys. 1957, 678, (26.
- R.J. Kern. Acrylonitrile and propionitrile complexes of metal chlorides in the first transition series. Journal of Inorganic and Nuclear Chemistry. 1963, 25, (1), 5-9.