Thermo Scientific Chemicals

Methyl 2-iodobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A13204.09
10 g, Each
Thermo Scientific Chemicals

Methyl 2-iodobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A13204.09
10 g, Each
Quantity
Catalog number: A13204.09
also known as A13204-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-97-9
IUPAC Name
methyl 2-iodobenzoate
Molecular Formula
C8H7IO2
InChI Key
BXXLTVBTDZXPTN-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1I
Form
Liquid
Refractive Index
1.6020-1.6060 @ 20?C
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless to yellow

Description

2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Insoluble in water.

Notes
Light sensitive. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text