Thermo Scientific Chemicals

endo-N-Hydroxy-5-norbornene-2,3-dicarboximide, 97%, Thermo Scientific Chemicals

Catalog number: A13205.30
250 g, Each
Thermo Scientific Chemicals

endo-N-Hydroxy-5-norbornene-2,3-dicarboximide, 97%, Thermo Scientific Chemicals

Catalog number: A13205.30
250 g, Each
Quantity
Catalog number: A13205.30
also known as A13205-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21715-90-2
IUPAC Name
4-hydroxy-4-azatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular Formula
C9H9NO3
InChI Key
ZUSSTQCWRDLYJA-UHFFFAOYNA-N
SMILES
ON1C(=O)C2C3CC(C=C3)C2C1=O
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to pale cream
Melting Point (clear melt)
163.0-173.0?C

Description

It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It decreases racemization in peptide synthesis and inhibits formation of N-acylureas.

Solubility
Soluble in ethanol

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text