Thermo Scientific Chemicals

4'-Aminoacetophenone, 99%, Thermo Scientific Chemicals

Catalog number: A13213.18
50 g, Each
Thermo Scientific Chemicals

4'-Aminoacetophenone, 99%, Thermo Scientific Chemicals

Catalog number: A13213.18
50 g, Each
Quantity
Catalog number: A13213.18
also known as A13213-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-92-3
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
Pale yellow to cream to brown
Assay (GC)
≥98.5%
Melting Point (clear melt)
103.0-109.0?C

Description

4'-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

Solubility
It is soluble in hot water, ethanol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. acids, Incompatible to Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents, Strong reducing agents. Stable under ordinary conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text