4'-Aminoacetophenone, 99%, Thermo Scientific Chemicals
4'-Aminoacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Aminoacetophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
50 g
1000 g
Catalog number A13213.30
also known as A13213-30
Price (USD)/ Each
137.65
Online exclusive
153.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
137.65
Online exclusive
153.00 
Save 15.35 (10%)
Add to cart
4'-Aminoacetophenone, 99%, Thermo Scientific Chemicals
Catalog numberA13213.30
Price (USD)/ Each
137.65
Online exclusive
153.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS99-92-3
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)Pale yellow to cream to brown
Assay (GC)≥98.5%
Melting Point (clear melt)103.0-109.0?C
4'-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Aminoacetophenone, is used in pharmaceuticals, medicines and other organic products. It is also used as a reagent for the photometric determination of Ce. As pharmaceutical intemediate or in the determination of palladium and vitaminB1.

Solubility
It is soluble in hot water, ethanol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. acids, Incompatible to Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents, Strong reducing agents. Stable under ordinary conditions.
RUO – Research Use Only

General References:

  1. Bushra Sulaiman.; Wadala A. Bashir. Spectrophotometric determination of 1-naphthylamine in aqueous solution by coupling with diazotised 4-aminoacetophenone. Analyst.1984, 109, (11), 1409-1411.
  2. Peter Wan.; Xigen Xu. Disproportionation of 4-nitroacetophenone to 4-aminoacetophenone and 4-nitrobenzoic acid. J. Org. Chem. 1989, 54, (18), 4473-4474.