Thermo Scientific™

3-Aminopyridine, 99%, Thermo Scientific Chemicals

Catalog number: A13216.30
250 g, Each
Thermo Scientific™

3-Aminopyridine, 99%, Thermo Scientific Chemicals

Catalog number: A13216.30
250 g, Each
Quantity
Catalog number: A13216.30
also known as A13216-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
462-08-8
IUPAC Name
pyridin-3-amine
Molecular Formula
C5H6N2
InChI Key
CUYKNJBYIJFRCU-UHFFFAOYSA-N
SMILES
NC1=CC=CN=C1
Melting Point (clear melt)
55-63°C
Appearance (Color)
White to cream to brown or orange
Form
Crystals or crystalline powder or lumps or fused solid or flakes
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms

Description

3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

Solubility
It is soluble in water as well as soluble in alcohol, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text