Thermo Scientific Chemicals

2-Bromo-4,6-di-tert-butylphenol, 97%, Thermo Scientific Chemicals

Catalog number: A13218.22
100 g, Each
Thermo Scientific Chemicals

2-Bromo-4,6-di-tert-butylphenol, 97%, Thermo Scientific Chemicals

Catalog number: A13218.22
100 g, Each
Quantity
Catalog number: A13218.22
also known as A13218-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20834-61-1
IUPAC Name
2-bromo-4,6-di-tert-butylphenol
Molecular Formula
C14H21BrO
InChI Key
DIWZVAHZEOFSLS-UHFFFAOYSA-N
SMILES
CC(C)(C)C1=CC(Br)=C(O)C(=C1)C(C)(C)C
Appearance (Color)
White to cream or yellow to orange
Assay (GC)
≥96.0%
Melting Point
54.5-59.5?C
Form
Crystals or powder or crystalline powder or lumps
Identification (FTIR)
Conforms

Description

It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in pharmaceutical intermediates and as medicine. When 2-Bromo-4,6-di-tert-butylphenol was treated with potassium ferricyanide in benzene 1,4-dihydro-4-bromo-2,4,6,8-tetra-tert-butyl-1-oxodibenzofuran was yielded with a small amount of 2,4,6,8-tetra-tert-butyldibenzofuran.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text