N-Phenylbenzamidine, 97%, Thermo Scientific Chemicals
N-Phenylbenzamidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Phenylbenzamidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13219.06
also known as A13219-06
Price (USD)/ Each
78.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
78.10
Add to cart
N-Phenylbenzamidine, 97%, Thermo Scientific Chemicals
Catalog numberA13219.06
Price (USD)/ Each
78.10
-
Add to cart
Chemical Identifiers
CAS1527-91-9
IUPAC Name[phenyl(phenylamino)methylidene]azanium
Molecular FormulaC13H13N2
InChI KeyMPYOKHFSBKUKPQ-UHFFFAOYSA-O
SMILES[NH2+]=C(NC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)110-120?C
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only