Thermo Scientific Chemicals

L-Phenylalanine, 99%

Catalog number: A13238.0B
1000 g, Each
Thermo Scientific Chemicals

L-Phenylalanine, 99%

Catalog number: A13238.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13238.0B
also known as A13238-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63-91-2
IUPAC Name
2-amino-3-phenylpropanoic acid
Molecular Formula
C9H11NO2
InChI Key
COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES
NC(CC1=CC=CC=C1)C(O)=O
Appearance (Color)
White
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Optical Rotation
-34 ? 1? (c=2 in Water)

Description

L-Phenylalanine is an aromatic organic compound containing amine and acid as functional groups. It is an essential amino acid and its structure can be considered as the derivative of 3-phenylpropionic acid, in which one hydrogen atom present at C-2 position is substituted with amino group. It is a chiral compound and consists of two optical isomers. It is also known as 2-amino-3-phenylpropanoic acid, beta-phenylalanine, L-alfa-amino-beta-phenylpropionic acid, H-Phe-OH and (S)-2-amino-3-phenylpropionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Phenylalanine is an essential amino acid found in several dietary sources, including meat, fish, eggs, cheese, and milk
  • Phenylalanine can be converted into the amino acid tyrosine

Application

  • L-Phenylalanine is used in the synthesis of other amino acids
  • It can be used as a precursor for the skin pigment melanin as well as the monoamine neurotransmitters dopamine, norepinephrine (noradrenaline), and epinephrine (adrenaline)
  • It is used to biochemically form proteins coded by DNA
  • Due to its ability to absorb ultraviolet radiation, it is used to quantify the amount of protein
  • In research, it is used as an alkaline phosphatase inhibitor
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text