Thermo Scientific Chemicals

Triphenylethylene, 98+%, Thermo Scientific Chemicals

Catalog number: A13244.30
250 g, Each
Thermo Scientific Chemicals

Triphenylethylene, 98+%, Thermo Scientific Chemicals

Catalog number: A13244.30
250 g, Each
Quantity
Catalog number: A13244.30
also known as A13244-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58-72-0
IUPAC Name
(2,2-diphenylethenyl)benzene
Molecular Formula
C20H16
InChI Key
MKYQPGPNVYRMHI-UHFFFAOYSA-N
SMILES
C(=C(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
65.0-71.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text