Thermo Scientific Chemicals

Propiolic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A13245.14
25 g, Each
Thermo Scientific Chemicals

Propiolic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A13245.14
25 g, Each
Quantity
Catalog number: A13245.14
also known as A13245-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-25-0
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Appearance (Color)
Clear, colorless to pale yellow or pale orange to brown
Refractive Index
1.4305-1.4345 @20?C
Form
Liquid

Description

Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.

Solubility
Miscible with water, ether, ethanol and chloroform.

Notes
Store in cool place. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text