Thermo Scientific Chemicals

2,4-Difluoro-1-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13249.22
100 g, Each
Thermo Scientific Chemicals

2,4-Difluoro-1-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A13249.22
100 g, Each
Quantity
Catalog number: A13249.22
also known as A13249-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
446-35-5
IUPAC Name
2,4-difluoro-1-nitrobenzene
Molecular Formula
C6H3F2NO2
InChI Key
RJXOVESYJFXCGI-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(F)C=C1F
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.5%
Refractive Index
1.5110-1.5150 @ 20°C
Form
Liquid
Identification (FTIR)
Conforms

Description

2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text