Invitrogen™

PrestoBlue™ Cell Viability Reagent

Catalog number: A13262
Have Questions?
Invitrogen™

PrestoBlue™ Cell Viability Reagent

Catalog number: A13262
Have Questions?
Catalog Number
A13262
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A13262100 mL
-
A1326125 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Get high quality cell viability results and instant time savings with PrestoBlue® Cell Viability Reagent. PrestoBlue® is a ready to use cell permeable resazurin-based solution that functions as a cell viability indicator by using the reducing power of living cells to quantitatively measure the proliferation of cells. When added to cells, the PrestoBlue® reagent is modified by the reducing environment of the viable cell and turns red in color, becoming highly fluorescent. This color change can be detected using fluorescence or absorbance measurements.

The PrestoBlue® Cell Viability Reagent allows you to:

• Save time with an incubation step as short as 10 minutes.
• Obtain high quality results with a large dynamic range
• Experience convenience with the add and read homogenous format
• Monitor living cells and perform downstream functional assays

10-minute incubations
The PrestoBlue® Cell Viability Reagent is the only live-cell viability reagent on the market able to provide robust data typically in as little as 10 minutes following reagent addition. All other resazurin-based cell viability reagents on the market require a one to four hour incubation with the reagent prior to obtaining data.

High quality results
Based on the cell type and incubation time, PrestoBlue® Cell Viability Reagent can detect as few as 12 cells per well in a 384 well plate. After ten minutes of incubation the PrestoBlue® Reagent can linearly detect cells down to 98 cells⁄well with a Z’ value of greater than 0.5.

Homogenous assay format
The PrestoBlue® Cell Viability Reagent is an addition-only reagent. Without the cell lysis or solubilization steps required by other assays on the market, hands-on time is minimized.

Live cell assays
The PrestoBlue® Cell Viability Reagent does not require cell lysis. Cells may be incubated for up to 24 hours in the presence of PrestoBlue® Cell Viability Product. Simply remove the reagent from the cells and replace it with growth medium for further proliferation.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Bacteria, Eukaryotic Cells, Fungal Cells
Color
Blue
Concentration
10 X
Description
PrestoBlue Cell Viability Reagent
Detection Method
Colorimetric, Fluorescence
For Use With (Application)
Viability Assay
For Use With (Equipment)
Microplate Reader, Spectrophotometer, HTS Reader
Product Type
Reagent
Dye Type
Resazurin
Emission
590-615 nm
Excitation Wavelength Range
535-560 nm
Form
Liquid
Format
96-well plate, Cuvettes, 384-well plate
Incubation Time
10 min. to 2 hr.
Product Line
PrestoBlue
Quantity
100 mL
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice
Throughput
High-throughput Compatible

Contents & Storage

Store at 4°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text