Thermo Scientific Chemicals

5-Methyl-2-furaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A13264.22
100 g, Each
Thermo Scientific Chemicals

5-Methyl-2-furaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: A13264.22
100 g, Each
Quantity
Catalog number: A13264.22
also known as A13264-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-02-0
IUPAC Name
5-methylfuran-2-carbaldehyde
Molecular Formula
C6H6O2
InChI Key
OUDFNZMQXZILJD-UHFFFAOYSA-N
SMILES
CC1=CC=C(O1)C=O
Refractive Index
1.5290-1.5330 @ 20?C
Form
Liquid
Free acid (titration)
≤1%
Appearance (Color)
Clear pale yellow to yellow to dark yellow to brown to dark brown
Assay (GC)
≥97.5%

Description

5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-2-furaldehyde is used as a perfuming agent. It is used a smoke with essence.

Solubility
Soluble in alcohol and water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text