Thermo Scientific Chemicals

Pyridazine, 98+%, Thermo Scientific Chemicals

Catalog number: A13266.06
5 g, Each
Thermo Scientific Chemicals

Pyridazine, 98+%, Thermo Scientific Chemicals

Catalog number: A13266.06
5 g, Each
Quantity
Catalog number: A13266.06
also known as A13266-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
289-80-5
IUPAC Name
pyridazine
Molecular Formula
C4H4N2
InChI Key
PBMFSQRYOILNGV-UHFFFAOYSA-N
SMILES
C1=CC=NN=C1
Appearance (Color)
Clear colorless to yellow to brown
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.5220-1.5260 @ 20?C

Description

Pyrrolo[1,2-b]pyridazine 1 is a N-bridgehead aromatic heterocycle containing two nitrogen atoms, is obtained by the condensation of pyridazine and pyrrole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrolo[1,2-b]pyridazine 1 is a N-bridgehead aromatic heterocycle containing two nitrogen atoms, is obtained by the condensation of pyridazine and pyrrole.

Solubility
Miscible with water, benzene, DMF. Soluble in alcohol, ether

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text