Thermo Scientific Chemicals

(+/-)-1-Octyn-3-ol, 98%, Thermo Scientific Chemicals

Catalog number: A13267.18
50 g, Each
Thermo Scientific Chemicals

(+/-)-1-Octyn-3-ol, 98%, Thermo Scientific Chemicals

Catalog number: A13267.18
50 g, Each
Quantity
Catalog number: A13267.18
also known as A13267-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
818-72-4
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4395-1.4435 @ 20?C
Form
Liquid
Assay (GC)
≥97.5%

Description

1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text