1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octyn-3-ol was used in the synthesis of synthetic tricolorin A which is an organic chemical.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
RUO – Research Use Only
Fabrizio Zucchi.; Giordano Trabanelli.; Giancarlo Brunoro. The influence of the chromium content on the inhibitive efficiency of some organic compounds.Corros. Sci. 1992, 33 (7),1135-1139 .
Gareth D. Rees.; Brian H. Robinson.; G.Richard Stephenson. Preparative-scale kinetic resolutions catalysed by microbial lipases immobilised in AOT-stabilised microemulsion-based organogels: cryoenzymology as a tool for improving enantioselectivity.Biochimica et Biophysica Acta (BBA) - Lipids and Lipid Metabolism. 1995, 1259 (1), 73-81 .
Selective monohydrobromination is described, using a combination of anhydrous HBr and tetraethylammonium bromide in dichloromethane, to give 2-bromo-1-octen-3-ol, as the first step in a synthesis of substituted furans: Org. Synth., 76, 263 (1998).