Thermo Scientific Chemicals

Bromopentafluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13273.14
25 g, Each
Thermo Scientific Chemicals

Bromopentafluorobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A13273.14
25 g, Each
Quantity
Catalog number: A13273.14
also known as A13273-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
344-04-7
IUPAC Name
1-bromo-2,3,4,5,6-pentafluorobenzene
Molecular Formula
C6BrF5
InChI Key
XEKTVXADUPBFOA-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(Br)C(F)=C1F
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.4480-1.4510 @ 20?C

Description

It is used as pharmaceutical intermediate. It is a Grignard reagent (with CuBr in dioxane) can introduce pentafluorophenyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. It is a Grignard reagent (with CuBr in dioxane) can introduce pentafluorophenyl groups.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text