Sodium 1-hexanesulfonate is used as a base in the organic coupling reactions such as Suzuki coupling reactions. It is used as an ion pairing reagent for HPLC and involved in the analysis of peptides and proteins. It is also used in high performance capillary electrophoresis analysis of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium 1-hexanesulfonate is used as a base in the organic coupling reactions such as Suzuki coupling reactions. It is used as an ion pairing reagent for HPLC and involved in the analysis of peptides and proteins. It is also used in high performance capillary electrophoresis analysis of peptides.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
Tsunoda, M. T. Analytical Methods for the Measurement of Catechol-O-Methyltransferase Activity in Animal Tissues. Curr. Biotechnol. 2015, 4 (2), 197-201.
Costa, A. L.; Gomes, A. C.; Pillinger, M.; Gonçalves, I. S.; de Melo, J. An Indigo Carmine-Based Hybrid Nanocomposite with Supramolecular Control of Dye Aggregation and Photobehavior. Chem. Eur. J. 2015, 21 (34), 12069-12078.