Thermo Scientific Chemicals

3-(Chloromethyl)pyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A13279.14
25 g, Each
Thermo Scientific Chemicals

3-(Chloromethyl)pyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A13279.14
25 g, Each
Quantity
Catalog number: A13279.14
also known as A13279-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6959-48-4
IUPAC Name
hydrogen 3-(chloromethyl)pyridine chloride
Molecular Formula
C6H7Cl2N
InChI Key
UZGLOGCJCWBBIV-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCC1=CC=CN=C1
Appearance (Color)
White to pale brown
Form
Crystalline powder or powder or needles
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Assay (Freebase GC)
≥96.0%
Water Content (Karl Fischer Titration)
<1.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text