Thermo Scientific Chemicals

Dimethyl sulfoxide, 99+%, Thermo Scientific Chemicals

Catalog number: A13280.36
500 g, Each
Thermo Scientific Chemicals

Dimethyl sulfoxide, 99+%, Thermo Scientific Chemicals

Catalog number: A13280.36
500 g, Each
Quantity
Catalog number: A13280.36
also known as A13280-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-68-5
IUPAC Name
methanesulfinylmethane
Molecular Formula
C2H6OS
InChI Key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES
CS(C)=O
Appearance (Color)
Clear colorless
Assay (GC)
≥99.0% (UK sourced material)
Water Content (Karl Fischer Titration)
≤0.5%
Assay from Supplier's CofA
≥99.0% (GC, US sourced material)
Refractive Index
1.4775-1.4805 @ 20?C (UK sourced material)

Description

Dimethyl Sulfoxide (DMSO) is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. It is also used in antifreeze or hydraulic fluids and utilized in the oxidation of thiols and disulfides to sulfonic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl Sulfoxide (DMSO) is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. It is also used in antifreeze or hydraulic fluids and utilized in the oxidation of thiols and disulfides to sulfonic acids. It is also used as a cryoprotectant and is added to cell media to reduce ice formation and thereby prevent cell death during the freezing process.

Solubility
Miscible with water, ethanol, ether, benzene, chloroform and acetone.

Notes
Hygroscopic. Incompatible with acid chlorides, phosphorus halides, strong acids, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text