2,4,5-Trihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals
2,4,5-Trihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,5-Trihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

2,4,5-Trihydroxybenzaldehyde, 97%, C7H6O4, CAS Number-35094-87-2, benzaldehyde, 2,4,5-trihydroxy, benzaldehyde,2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxybenzaldehyde, 2,4,5-trihydroxy-benzaldehyde, 5g, 2250084, 154.12, H315-H319-H335
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A13283.03
also known as A13283-03
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
2,4,5-Trihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberA13283.03
Price (USD)/ Each
164.00
-
Add to cart
Chemical Identifiers
CAS35094-87-2
IUPAC Name2,4,5-trihydroxybenzaldehyde
Molecular FormulaC7H6O4
InChI KeyWNCNWLVQSHZVKV-UHFFFAOYSA-N
SMILESOC1=CC(O)=C(C=O)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to brown to yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only