1x1 image pixel for data collection
GeneArt™ Seamless Cloning and Assembly Kit
Product Image
Invitrogen™
GeneArt™ Seamless Cloning and Assembly Kit
The GeneArt™ Seamless Cloning and Assembly Kit enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consistingRead more
Have Questions?
Catalog NumberQuantity
A1328820 Reactions
Catalog number A13288
Price (USD)
705.65
Online Exclusive
772.00
Save 66.35 (9%)
Each
Add to cart
Quantity:
20 Reactions
Price (USD)
705.65
Online Exclusive
772.00
Save 66.35 (9%)
Each
Add to cart
The GeneArt™ Seamless Cloning and Assembly Kit enables the simultaneous and directional cloning of 1 to 4 PCR fragments, consisting of any sequence, into any linearized vector, in a single 30-minute room temperature reaction. The kit contains everything required for the assembly of DNA fragments, and their transformation into E. coli for selection and growth of recombinant vectors.
Speed and Ease — Clone up to 4 DNA fragments, with sequence of your choice, simultaneously in a single vector (up to 13 Kb); no restriction digestion, ligation or recombination sites required
Precision and Efficiency —Designed to let you clone what you want, where you want, in the orientation you want, and achieve up to 90% correct clones with no extra sequences left behind
Vector Flexibility — Use our linear vector or a vector of your choice
Free Tools — Design DNA oligos and more with our free web-based interface that walks you step-by-step through your project
Diverse Applications — Streamline many synthetic biology and molecular biology techniques through the rapid combination, addition, deletion, or exchange of DNA segments
For cloning more than 4 DNA fragments, final molecules larger than 110 Kb, or for using pre-existing DNA elements too long to be amplified by PCR; please consider the GeneArt™ High-Order Genetic Assembly System (cat# A13285).
Simple and Fast Clone Creation
GeneArt™ Seamless Cloning is a simple, two step process, consisting of a tube based assembly reaction followed by transformation into One Shot™ Chemically Competent TOP10 E. coli. The kit uses the properties of a proprietary enzymatic mix to assemble DNA fragments with shared terminal end homology without leaving any extra sequences or scars behind (seamless). A portion of the assembly reaction is then transformed into the provided competent TOP10 cells, yielding clones ready for analysis the next day. The required 15-base pair end-homology can be easily engineered by PCR-amplification with custom DNA oligos.
Cloning Efficiency, Flexibility, and Precision
With the GeneArt™ Seamless Cloning and Assembly Kit the main factors effecting cloning efficiency are the size of the DNA elements (100 bp to 5 Kb), the total size of the final molecule (≤ 13 Kb), and the quality and specificity of the fragment. The terminal end of the PCR fragments (A-overhangs or blunt), does not affect the cloning efficiency.
Typical cloning efficiencies for different numbers of fragments cloned into pUC19L are the following:
• 90% for a single 5 Kb DNA element
• 70% for 4 fragments of 1 Kb each
• 40% for 4 DNA fragments of 2 Kb each
Success of the cloning is independent of the insert sequence and vector type, allowing you to design and add nearly any desired sequence, or combination of sequences, to any plasmid as long as it can be linearized by either restriction enzyme digestion or by PCR. The circularized clones obtain from the reaction contain only the sequence of your original vector, inserts, and designated homologies, with no extraneous nucleotide insertions.
in silico Cloning Design Support
A key step in GeneArt™ Seamless Cloning is the correct design of fragments and oligos with the appropriate homology and spacing to help ensure successful assembly of your clone. To simplify and speed the design process we provide a free online tool to help you design your experiment in silico. The tool checks for compatibility of the experimental design with the product specifications, designs DNA oligos with end-homology for the PCR amplification of the different elements to clone, and presents the user with a graphical representation of the vector, as well as a downloadable annotated sequence in GenBank format that is compatible with Vector NTI™ software.
Applications
The GeneArt™ Seamless Cloning and Assembly Kit is designed to empower cloning and DNA assembly experiments in a wide range of molecular biology and synthetic biology applications, among others. The product allows for the creation of modular expression vectors, with interchangeable parts, and can be used to perform a variety of tasks that would otherwise involve multiple steps. Use the kit to simply: construct fusion proteins, delete, replace, or add DNA elements such as restriction sites in an existing vector, and many other techniques that require manipulation of genetic sequences.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial or Yeast StrainTOP10
Cell TypeChemically Competent E. coli
Cloning MethodSeamless Cloning
For Use With (Application)Cloning
Number of FragmentsUp to 4 Fragments
Product LineGeneArt™
Product TypeCloning and Assembly Kit
Quantity20 Reactions
Size13 kb total (vector plus all inserts)
VectorpUC19L Linearized
Workflow StepDNA Assembly
FormatKit
Unit SizeEach
Contents & Storage
The kit has enough reagents for 20 cloning reactions and a control. The components are: 10X Enzyme Mix and 5X Reaction Buffer, control vector and insert, One Shot™ Chemically Competent TOP10 E. coli, S.O.C. Medium, and a supercoiled control plasmid for E.coli.
A cloning vector is not included in this kit.
For your convenience and flexibility, a ready-to-use linear vector is available separately for 20 cloning reactions (GeneArt™ Linear pUC19L Vector for Seamless Cloning, catalog number A13289).
Frequently asked questions (FAQs)
In the GeneArt Seamless Cloning and Assembly kit, is there a size limit for the 4 individual fragments that I can clone into the vector?
There is no size limit on the individual fragments as long as the combined total length of the 4 individual fragments and vector does not exceed 13 kb.
With the GeneArt Seamless Cloning and Assembly Kit, a large number of the transformants contain the incorrect insert. Can you please offer some suggestions?
Check your PCR product on a gel. You may be getting multiple products, causing incorrect inserts to clone in to your vector. Gel purification of the PCR products can help with this issue.
With the GeneArt Seamless Cloning and Assembly Kit, a large number of the transformants contain no insert. Can you please offer some suggestions?
Check to ensure that the cloning vector is completely linearized. Additionally, the order in which the GeneArt Enzyme mix is added is crucial to the experiment – add it last. Lastly, check the incubation time of the reaction for the recommended time.
With the GeneArt Seamless Cloning and Assembly Kit, I'm getting no colonies after transformation with DNA inserts, but the transformation with control assembly reaction is successful. Can you please offer some troubleshooting tips?
– Check the purity of the PCR products.
– Ensure that the required end-terminal homology between ends is present.
– DNA ends may have been damaged during preparation (exposure to UV); limit exposure time to UV/EtBr.
– Check ratio and amounts of DNA inserts and vector (2:1 insert:vector molar ratio).
– Ensure that the GeneArt Enzyme Mix is handled correctly.This enzyme is temperature sensitive. Return immediately to storage after use. Do not leave at room temperature or on ice for extended periods.
I used electrocompetent cells instead of chemically competent cells with the GeneArt Seamless Cloning and Assembly Kit and now my reaction isn’t working. Why is this?
We do not recommend using electrocompetent cells. The enzyme mix does not perform ligation of the DNA ends, so electroporation will disrupt the DNA base pairs formed during assembly.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link