Thermo Scientific Chemicals

Tetramethylammonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: A13288.0I
5000 g, Each
Thermo Scientific Chemicals

Tetramethylammonium chloride, 97%, Thermo Scientific Chemicals

Catalog number: A13288.0I
5000 g, Each
Quantity
Catalog number: A13288.0I
also known as A13288-0I
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-57-0
IUPAC Name
tetramethylazanium chloride
Molecular Formula
C4H12ClN
InChI Key
OKIZCWYLBDKLSU-UHFFFAOYSA-M
SMILES
[Cl-].C[N+](C)(C)C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤1.0%

Description

Tetramethylammonium binds AT-rich DNA polymers while concomitantly abolishing the preferential melting of AT versus GC base pairs. Supplied as 0.2 ?m filtered solution in 18 megohm water. It is employed as a phase transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium binds AT-rich DNA polymers while concomitantly abolishing the preferential melting of AT versus GC base pairs. Supplied as 0.2 μm filtered solution in 18 megohm water. It is employed as a phase transfer catalyst.

Solubility
Soluble in water. Insoluble in ether, and benzene.

Notes
Hygroscopic. Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text