Thermo Scientific Chemicals

Dodecanoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13294.22
100 g, Each
Thermo Scientific Chemicals

Dodecanoyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A13294.22
100 g, Each
Quantity
Catalog number: A13294.22
also known as A13294-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-16-3
IUPAC Name
dodecanoyl chloride
Molecular Formula
C12H23ClO
InChI Key
NQGIJDNPUZEBRU-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCC(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4440-1.4480 @ 20?C

Description

Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

Solubility
Soluble in ethanol and methanol.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text