Dodecanoyl chloride, 98%, Thermo Scientific Chemicals
Dodecanoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dodecanoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number A13294.36
also known as A13294-36
Price (USD)/ Each
133.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
133.00
Add to cart
Dodecanoyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA13294.36
Price (USD)/ Each
133.00
-
Add to cart
Chemical Identifiers
CAS112-16-3
IUPAC Namedodecanoyl chloride
Molecular FormulaC12H23ClO
InChI KeyNQGIJDNPUZEBRU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4440-1.4480 @ 20?C
Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

Solubility
Soluble in ethanol and methanol.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Shoji Fujii.; Hidenobu Kumagai.; Manjiro Noda. Preparation of poly(acyl)chitosans. Carbohydr. Res. 1980, 83 (2),389-393 .
  2. Daniel Pasquini.; Mohamed Naceur Belgacem.; Alessandro Gandini.; Antonio Aprigio da Silva Curvelo. Surface esterification of cellulose fibers: Characterization by DRIFT and contact angle measurements. J. Colloid Interface Sci. 2006, 295 (1),79-83 .