Thermo Scientific Chemicals

4-Methylbenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: A13297.22
100 g, Each
Thermo Scientific Chemicals

4-Methylbenzyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: A13297.22
100 g, Each
Quantity
Catalog number: A13297.22
also known as A13297-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-81-4
IUPAC Name
1-(bromomethyl)-4-methylbenzene
Molecular Formula
C8H9Br
InChI Key
WZRKSPFYXUXINF-UHFFFAOYSA-N
SMILES
CC1=CC=C(CBr)C=C1
Appearance (Color)
White to pale cream to pale brown
Form
Crystals or powder or crystalline powder or fused solid
Identification (FTIR)
Conforms
Melting Point (clear melt)
32.0-38.0?C
Assay (GC)
≥97.5%

Description

4-Methylbenzyl bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzyl bromide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text