Thermo Scientific Chemicals

Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals

Catalog number: A13298.30
250 g, Each
Thermo Scientific Chemicals

Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals

Catalog number: A13298.30
250 g, Each
Quantity
Catalog number: A13298.30
also known as A13298-30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
688-73-3
IUPAC Name
tributylstannyl
Molecular Formula
C12H27Sn
InChI Key
PIILXFBHQILWPS-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)CCCC
Form
Liquid
Refractive Index
1.4705-1.4755 @ 20?C
Comment
May form slight precipitate on storage. May develop pressure.
Appearance (Color)
Clear or slightly hazy, colorless
Assay (GC)
≥96.0%

Description

Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin hydride is used as a radical reagent in reductive cleavage, radical dehalogenation and intramolecular radical cyclization. It also promotes intramolecular cyclization leading to isoxazolo-benzaulene ring system. Further, it acts as a reducing agent for the conversion of alkyl halides to hydrocarbons. In addition to this, it is used in a veterinary anthelmintic for poultry and an intermediate to make other butyltin compounds.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text