Thermo Scientific Chemicals

Phthaldialdehyde, 98%, Thermo Scientific Chemicals

Catalog number: A13299.18
50 g, Each
Thermo Scientific Chemicals

Phthaldialdehyde, 98%, Thermo Scientific Chemicals

Catalog number: A13299.18
50 g, Each
Quantity
Catalog number: A13299.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
643-79-8
IUPAC Name
benzene-1,2-dicarbaldehyde
Molecular Formula
C8H6O2
InChI Key
ZWLUXSQADUDCSB-UHFFFAOYSA-N
SMILES
O=CC1=CC=CC=C1C=O
Assay (GC)
≥97.5
Identification (FTIR)
Conforms
Form
Crystals, crystalline powder or lumps
Free acid (titration)
≤1.0% (as Phthalic acid)
Melting Point (clear melt)
51-59?C

Description

Phthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthaldialdehyde is used as a fluorescent reagent for checking the assay of amines present in proteins, peptides by capillary electrophoresis and chromatography. It is also used for cholesterol assay in the picomole range as well as for fluorometric determination of histamine, histidine and other amino acids. Poly(phthalaldehyde) is obtained by the polymerization reaction which is used to make a photoresist. The nitrogen by o-phthaldialdehyde assay (NOPA) is a method used in winemaking process to measure the completion of fermentation.

Solubility
Soluble in acetic acid and methanol. Slightly soluble in water.

Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, amines and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text