Thermo Scientific Chemicals

Trimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals

Catalog number: A13301.14
25 g, Each
Thermo Scientific Chemicals

Trimethyl phosphonoacetate, 98%, Thermo Scientific Chemicals

Catalog number: A13301.14
25 g, Each
Quantity
Catalog number: A13301.14
also known as A13301-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5927-18-4
IUPAC Name
methyl 2-(dimethoxyphosphoryl)acetate
Molecular Formula
C5H11O5P
InChI Key
SIGOIUCRXKUEIG-UHFFFAOYSA-N
SMILES
COC(=O)CP(=O)(OC)OC
Appearance (Color)
Clear, colourless
Assay (GC)
>97.5%
Refractive Index
1.4350-1.4390 @ 20*C
Form
liquid

Description

Trimethyl phosphonoacetate is used in the preparation of sarin A by intramolecular Mannich-type reactions. It is involved in the Horner-Wadsworth-Emmons olefination with aldehydes and ketones to give acrylic esters. It is also involved in oxa-Michael reactions, prenylation of oxindoles and heterocyclization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl phosphonoacetate is used in the preparation of sarin A by intramolecular Mannich-type reactions. It is involved in the Horner-Wadsworth-Emmons olefination with aldehydes and ketones to give acrylic esters. It is also involved in oxa-Michael reactions, prenylation of oxindoles and heterocyclization reactions.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text