Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.
Solubility
Soluble in methanol.
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only
A. Cecinato.; V. Di Palo.; R. Mabili.; M. Possanzini. Pentafluorophenylhydrazine as a coating reagent for the HRGC-MS determination of semi-volatile carbonyl compounds in air.Chromatographia. 2001, 54 (3-4), 263-269 .
J. M. Birchall.; R. N. Haszeldine.; A. R. Parkinson. 966. Polyfluoroarenes. Part V. Pentafluorophenylhydrazine and the pentafluorophenyl radical.J. Chem. Soc. 1962, 4966-4976 .
Reagent for the derivatization of ketones for GC electron-capture detection: Anal. Biochem, 20, 394 (1967).