Thermo Scientific Chemicals

Pentafluorophenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: A13302.14
25 g, Each
Thermo Scientific Chemicals

Pentafluorophenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: A13302.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A13302.14
also known as A13302-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
828-73-9
IUPAC Name
(2,3,4,5,6-pentafluorophenyl)hydrazine
Molecular Formula
C6H3F5N2
InChI Key
BYCUWCJUPSUFBX-UHFFFAOYSA-N
SMILES
NNC1=C(F)C(F)=C(F)C(F)=C1F
Appearance (Color)
Pale cream to cream to brown
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or flakes

Description

Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorophenylhydrazine used to determine the purity of N-tert-butyl-N-[1-diethylphosphono-(2,2-dimethylpropyl)]nitroxide (DEPN) using 1H NMR spectroscopy.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text