Thermo Scientific Chemicals

3-Bromobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A13317.09
10 g, Each
Thermo Scientific Chemicals

3-Bromobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: A13317.09
10 g, Each
Quantity
Catalog number: A13317.09
also known as A13317-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6952-59-6
IUPAC Name
3-bromobenzonitrile
Molecular Formula
C7H4BrN
InChI Key
STXAVEHFKAXGOX-UHFFFAOYSA-N
SMILES
BrC1=CC=CC(=C1)C#N
Form
Crystals, crystalline powder or fused solid
Clarity
(2.5% w/v in Ethanol) : Transparent by JIS K8001 5.2
Appearance (Color)
White to pale yellow
Identification (FTIR)
Conforms
Assay (GC)
≥98.5%

Description

3-Bromobenzonitrile is involved in the Suzuki reaction under solvent-free conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobenzonitrile is involved in the Suzuki reaction under solvent-free conditions.

Solubility
Soluble in water (0.2 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text