Thermo Scientific Chemicals

3,5-Dichlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13325.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dichlorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: A13325.06
5 g, Each
Quantity
Catalog number: A13325.06
also known as A13325-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10203-08-4
IUPAC Name
3,5-dichlorobenzaldehyde
Molecular Formula
C7H4Cl2O
InChI Key
CASRSOJWLARCRX-UHFFFAOYSA-N
SMILES
ClC1=CC(C=O)=CC(Cl)=C1
Form
Crystals or powder or crystalline powder or fused solid
Free acid (titration)
≤1.5%
Appearance (Color)
White to cream
Assay (GC)
≥96.0%
Melting Point (clear melt)
57.5-66.5?C

Description

3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorobenzaldehyde was used as starting reagent during synthesis of β-aryl-β-amino acid enantiomers. It was used in the synthesis of dichlorophenylpyruvic acid.

Solubility
Insoluble in water.

Notes
Air sensitive. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text