Thermo Scientific Chemicals

Methanesulfonic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: A13328.14
25 g, Each
Thermo Scientific Chemicals

Methanesulfonic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: A13328.14
25 g, Each
Quantity
Catalog number: A13328.14
also known as A13328-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7143-01-3
IUPAC Name
methanesulfonyl methanesulfonate
Molecular Formula
C2H6O5S2
InChI Key
IZDROVVXIHRYMH-UHFFFAOYSA-N
SMILES
CS(=O)(=O)OS(C)(=O)=O
Appearance (Color)
White to grey to brown
Identification (FTIR)
Conforms
Form
Powder or crystals and/or chunks
Assay (Titration ex Anhydride)
≥96.0 to ≤104.0%
Melting Point (clear melt)
58-72°C

Description

Methanesulfonic anhydride is used to prepare methylsulfonyl esters called as mesylates. It finds application in the reaction of aromatic alkylation. It reacts with dimethylsulfoxide to get sulfoxonium complex, which is a useful oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methanesulfonic anhydride is used to prepare methylsulfonyl esters called as mesylates. It finds application in the reaction of aromatic alkylation. It reacts with dimethylsulfoxide to get sulfoxonium complex, which is a useful oxidizing agent.

Solubility
Soluble in organic solvents.

Notes
Moisture sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text