Thermo Scientific Chemicals

2,4-Dichlorobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A13338.14
25 g, Each
Thermo Scientific Chemicals

2,4-Dichlorobenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: A13338.14
25 g, Each
Quantity
Catalog number: A13338.14
also known as A13338-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1777-82-8
IUPAC Name
(2,4-dichlorophenyl)methanol
Molecular Formula
C7H6Cl2O
InChI Key
DBHODFSFBXJZNY-UHFFFAOYSA-N
SMILES
OCC1=CC=C(Cl)C=C1Cl
Appearance (Color)
White
Assay (GC)
≥98.5%
Form
Crystals or powder or crystalline powder or needles or lumpy crystalline powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
56.0-62.0°C

Description

2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

Solubility
Soluble in water, methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text