Thermo Scientific Chemicals

4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13347.03
1 g, Each
Thermo Scientific Chemicals

4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: A13347.03
1 g, Each
Quantity
Catalog number: A13347.03
also known as A13347-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5720-05-8
IUPAC Name
(4-methylphenyl)boronic acid
Molecular Formula
C7H9BO2
InChI Key
BIWQNIMLAISTBV-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)B(O)O
Assay (Aqueous acid-base Titration)
≥97.5%
Proton NMR
Conforms to structure
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale cream to cream
Assay (HPLC)
≥97.5%

Description

4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text