4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A13347.06
also known as A13347-06
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
Add to cart
4-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberA13347.06
Price (USD)/ Each
117.65
Online exclusive
130.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5720-05-8
IUPAC Name(4-methylphenyl)boronic acid
Molecular FormulaC7H9BO2
InChI KeyBIWQNIMLAISTBV-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5%
Proton NMRConforms to structure
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream
Assay (HPLC)≥97.5%
4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzeneboronic acid, is a commonly used chemical in Suzuki coupling reactions. It is also used as an intermediate in pharmaceutical industry

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Z Jia.; JW Quail.; VK Arora.; JR Dimmock. Structure of 3, 5-bis (benzylidene)-1-methyl-4-piperidone methobromide hemiethanol solvate. Tetrahedron Letters. 2001, 42,(29) 4751-4754.
  2. SD Taylor.;CC Kotoris.; AN Dinaut.; Q Wang. Potent non-peptidyl inhibitors of protein tyrosine phosphatase 1B. Bioorganic & Medicinal Chemistry. 1998, 6,(9), 1457-1468.
  3. GC reagent for diols: J. Chromat., 158, 33 (1978); 186, 307 (1979).
  4. For general chemistry of boronic acids, see Appendix 5.