Thermo Scientific Chemicals

Phosphorus(V) oxide, 98%, Thermo Scientific Chemicals

Catalog number: A13348.36
500 g, Each
Thermo Scientific Chemicals

Phosphorus(V) oxide, 98%, Thermo Scientific Chemicals

Catalog number: A13348.36
500 g, Each
Quantity
Catalog number: A13348.36
also known as A13348-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-56-3
IUPAC Name
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Molecular Formula
O5P2
InChI Key
DLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILES
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

Phosphorus(V) oxide is used as a drying and dehydrating agent, a condensation reagent in organic synthesis and a laboratory reagent. It is also used in sugar refining and in fire extinguishing. Phosphorus pentoxide in DMSO forms an Onodera reagent which oxidizes alcohols. It is capable of converting mineral acids to anhydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphorus(V) oxide is used as a drying and dehydrating agent, a condensation reagent in organic synthesis and a laboratory reagent. It is also used in sugar refining and in fire extinguishing. Phosphorus pentoxide in DMSO forms an Onodera reagent which oxidizes alcohols. It is capable of converting mineral acids to anhydrides.

Solubility
Soluble in sulfuric acid. Insoluble in acetone and ammonia. Decomposes in water.

Notes
It is highly hygroscopic in nature and gets hydrolyzed to phosphoric acid. It is stored in a separate safety cabinet away from incompatibles such as alkalis and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text