4-Hydroxy-3-methoxy-DL-mandelic acid, 98%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxy-DL-mandelic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-3-methoxy-DL-mandelic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A13349.06
also known as A13349-06
Price (USD)/ Each
162.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
162.00
Add to cart
4-Hydroxy-3-methoxy-DL-mandelic acid, 98%, Thermo Scientific Chemicals
Catalog numberA13349.06
Price (USD)/ Each
162.00
-
Add to cart
Chemical Identifiers
CAS55-10-7
IUPAC Name(2S)-2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetate
Molecular FormulaC9H9O5
InChI KeyCGQCWMIAEPEHNQ-QMMMGPOBSA-M
SMILESCOC1=CC(=CC=C1O)[C@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)129.0-135.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
Appearance (Color)Pale brown to brown to pale gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only