It can be used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used in organic synthesis.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only
Samuel R. Aspinall; Ernest J. Bianco. A Synthesis of 2-Alkylamino-4,5-dihydroimidazoles. J. Am. Chem. Soc,. 1951, 73 (2), 602-603.
Kiyoshi Tanemura; Tsuneo Suzuki; Yoko Nishida; Koko Satsumabayashi; Takaaki Horaguchi. Halogenation of Aromatic Compounds by N-chloro-, N-bromo-, and N-iodosuccinimide. Chemistry Letters, 2003, 32 (10), 932-933.