Thermo Scientific Chemicals

3,4-Dibromotoluene, 98+%, Thermo Scientific Chemicals

Catalog number: A13351.22
100 g, Each
Thermo Scientific Chemicals

3,4-Dibromotoluene, 98+%, Thermo Scientific Chemicals

Catalog number: A13351.22
100 g, Each
Quantity
Catalog number: A13351.22
also known as A13351-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60956-23-2
IUPAC Name
1,2-dibromo-4-methylbenzene
Molecular Formula
C7H6Br2
InChI Key
LDCPXNOCWDGYIU-UHFFFAOYSA-N
SMILES
CC1=CC=C(Br)C(Br)=C1
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Form
Liquid
Refractive Index
1.5980-1.6020 @ 20?C

Description

It can be used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text