Thermo Scientific Chemicals

1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals

Catalog number: A13352.0B
1000 g, Each
Thermo Scientific Chemicals

1-Methyl-4-piperidone, 98%, Thermo Scientific Chemicals

Catalog number: A13352.0B
1000 g, Each
Quantity
Catalog number: A13352.0B
also known as A13352-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1445-73-4
IUPAC Name
1-methylpiperidin-4-one
Molecular Formula
C6H11NO
InChI Key
HUUPVABNAQUEJW-UHFFFAOYSA-N
SMILES
CN1CCC(=O)CC1
Form
Liquid
Comment
May form precipitate on storage.
Appearance (Color)
Clear colorless to yellow to orange or yellow to brown
Assay (GC)
>97.5%
Refractive Index
1.4590-1.4630 @ 20?C

Description

1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-4-piperidone, are widely used in artificial fibre industry. They are polymerizable and used as nylon precursors. They have excellent applications as solvents and as intermediates for organic synthesis. They are used in the aqueous carrier medium and amide penetrants in inks and water soluble paints.

Solubility
It is miscible with water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text