Thermo Scientific Chemicals

4-Hydrazinobenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13362.14
25 g, Each
Thermo Scientific Chemicals

4-Hydrazinobenzenesulfonic acid hydrate, 98%, Thermo Scientific Chemicals

Catalog number: A13362.14
25 g, Each
Quantity
Catalog number: A13362.14
also known as A13362-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-71-5
IUPAC Name
4-hydrazinylbenzene-1-sulfonic acid
Molecular Formula
C6H8N2O3S
InChI Key
IOMZCWUHFGMSEJ-UHFFFAOYSA-N
SMILES
NNC1=CC=C(C=C1)S(O)(=O)=O
Appearance (Color)
Pale cream to cream to pale brown
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (HPLC)
≥97.5%
Identification (FTIR)
Conforms

Description

4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

Solubility
It is soluble in water.

Notes
Stable under normal conditions.Keep away from strong oxidizing agents. Store in closed vessels.Hygroscopic
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text